Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

857450P

Avanti

C6-Pyridinium-Ceramide

D-erythro-2-N-[6′-(1"-pyridinium)-hexanoyl]-sphingosine bromide, powder

Synonym(s):

D-erythro-C6-pyridinium-ceramide; LCL29

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H51N2O3Br
CAS Number:
Molecular Weight:
555.63
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857450P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857450P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

C6-Pyridinium-Ceramide is a water-soluble, cationic pyridinium analog of ceramide.[1] Structurally, ceramides have a sphingosine base and amide-linked acyl chains of different length.[2]

Application

C6-Pyridinium-Ceramide has been used in treating HeLa cell lines, to elucidate its role in alternative splicing and as a standard for high performance liquid chromatography (HPLC).[3] It has also been used in combination with photodynamic therapy for head and neck squamous cancer cell carcinoma[4] and mouse squamous cell carcinoma.[5]

Biochem/physiol Actions

C6-Pyridinium-Ceramide plays an important role in alternative splicing of the gene by inhibiting protein phosphatase-1.[3] C6-Pyridinium-Ceramide (LCL29) acts as an anti-cancer agent. It sensitizes cancer cells when combined with photodynamic therapy.[4] In cancer cells, LCL29 accumulates in the negatively charged mitochondria.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857450P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergei A Novgorodov et al.
The Journal of biological chemistry, 280(16), 16096-16105 (2005-02-22)
Ceramide-induced cell death is thought to be mediated by change in mitochondrial function, although the precise mechanism is unclear. Proposed models suggest that ceramide induces cell death through interaction with latent binding sites on the outer or inner mitochondrial membranes
C6-pyridinium ceramide sensitizes SCC17B human head and neck squamous cell carcinoma cells to photodynamic therapy
Boppana NB, et al.
Journal of Photochemistry and Photobiology. B, Biology, 143, 163-168 (2015)
Enhanced tumor cures after Foscan photodynamic therapy combined with the ceramide analog LCL29. Evidence from mouse squamous cell carcinomas for sphingolipids as biomarkers of treatment response
Separovic D, et al.
International journal of oncology, 38(2), 521-527 (2011)
Zdzislaw M Szulc et al.
Bioorganic & medicinal chemistry, 14(21), 7083-7104 (2006-08-22)
In the course of our studies on compartment-specific lipid-mediated cell regulation, we identified an intimate connection between ceramides (Cers) and the mitochondria-dependent death-signaling pathways. Here, we report on a new class of cationic Cer mimics, dubbed ceramidoids, designed to act
C6 pyridinium ceramide influences alternative pre-mRNA splicing by inhibiting protein phosphatase-1
Sumanasekera C, et al.
Nucleic Acids Research, 40(9), 4025-4039 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service