Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

857381P

Avanti

SLM6031434

Avanti Research - A Croda Brand (857381P), powder

Synonym(s):

(R)-2-(3-(4-(octyloxy)-3-(trifluoromethyl)phenyl)-1,2,4-oxadiazol-5-yl)pyrrolidine-1-carboximidamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30F3N5O2
CAS Number:
Molecular Weight:
453.50
UNSPSC Code:
12352211
NACRES:
NA.25

assay

99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (857381P-25mg)
pkg of 1 × 5 mg (857381P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand (857381P)

shipped in

dry ice

storage temp.

−20°C

General description

SLM6031434 is an inhibitor of sphingosine kinase type 2 (SphK2). It consists of pedant 1-guanidino-2-phenyloxadiazolylpyrrolidine group.

Biochem/physiol Actions

SLM6031434 increases the blood sphingosine 1-phosphate (S1P) levels in mice and this occurs due to the inhibition of sphingosine kinase type 2 (SphK2).

Packaging

5 mL Amber Glass Screw Cap Vial (857381P-25mg)
5 mL Amber Glass Screw Cap Vial (857381P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoacoustic microscopy reveals the hemodynamic basis of sphingosine 1-phosphate-induced neuroprotection against ischemic stroke
Cao R, et al.
Theranostics, 8(22), 6111-6111 (2018)
Saccharomyces cerevisiae as a platform for assessing sphingolipid lipid kinase inhibitors
Kharel Y, et al.
Testing, 13(4), 6111-6111 (2018)
Sphingosine kinase 2 inhibition and blood sphingosine 1-phosphate levels
Kharel Y, et al.
Journal of Pharmacology and Experimental Therapeutics, 355(1), 23-31 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service