Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

857380P

Avanti

 SLP7111228

Avanti Research - A Croda Brand, powder

Synonym(s):

(S)-2-((3-(4-octylphenyl)-1,2,4-oxadiazol-5-yl)methyl)pyrrolidine-1-carboximidamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H34ClN5O
CAS Number:
Molecular Weight:
419.99
UNSPSC Code:
12352211

assay

99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (857380P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

N=C(N)N1CCC[C@H]1CC2=NC(C3=CC=C(CCCCCCCC)C=C3)=NO2.Cl

General description

SLP7111228 is a selective inhibitor of sphingosine kinase type 1 (SphK1).[1] It consists of a pedant 1-guanidino-2-phenyloxadiazolylpyrrolidine group.[2]

Biochem/physiol Actions

SLP7111228 might lower the circulating levels of sphingosine 1-phosphate (S1P).[1]

Packaging

5ML Glass Amber Screw Cap Vial (857380P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosine kinase inhibitors: a review of patent literature (2006-2015)
Lynch K R, et al.
Expert Opinion on Therapeutic Patents, 26(12), 1409-1416 (2016)
Saccharomyces cerevisiae as a platform for assessing sphingolipid lipid kinase inhibitors
Kharel Y, et al.
Testing, 13(4) (2018)
Saccharomyces cerevisiae as a platform for assessing sphingolipid lipid kinase inhibitors
Kharel Y, et al.
PLoS ONE, 13(4) (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service