Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

857367P

Avanti

VPC 23153

Avanti Research - A Croda Brand 857367P, powder

Synonym(s):

(R)-phosphoric acid mono-[2-amino-2-(6-octyl-1H-benzoimiazol-2-yl)-ethyl] ester

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H28N3O4P
CAS Number:
Molecular Weight:
369.40
UNSPSC Code:
12352211

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857367P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857367P

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCC1=CC=C2C(N=C([C@H](COP(O)([O-])=O)[NH3+])N2)=C1

Application

VPC 23153 has been used to study its effects in isolated pulmonary, renal and mesenteric arteries.[1] It has also been used to study its pharmacological properties at the human sphingosine 1-phosphate 3 (S1P3)receptor expressed in Chinese hamster ovary (CHO) FlpIn cells.[2]

Biochem/physiol Actions

VPC 23153 is a sphingosine 1-phosphate 4 (S1P4) receptor agonist. It is associated with the contraction of pulmonary arteries in rats.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857367P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Ota et al.
Pulmonary circulation, 1(3), 399-404 (2011-12-06)
This study aimed to identify receptors mediating sphingosine-1-phosphate (S1P)-induced vasoconstriction in the normotensive and chronic hypoxia-induced hypertensive rat pulmonary circulation. In isolated perfused lungs from normoxic rats, infusion of S1P caused a sustained vasoconstriction, which was not reduced by combinational
M Jongsma et al.
British journal of pharmacology, 156(8), 1305-1311 (2009-03-25)
Recently, some ligands targeting the sphingosine-1-phosphate receptor subtype 3 (S1P(3)) have become available. The characterization of these compounds was mainly based on one functional read-out system, although S1P(3) receptors are known to activate different signal transduction pathways. Therefore, this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service