Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

857353P

Avanti

VPC 31143 (R)

Avanti Research - A Croda Brand 857353P, powder

Synonym(s):

N-{(1R)-2-hydroxy-1-[(phosphonooxy)methyl]ethyl}(9Z)octadec-9-enamide (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H45N2O6P
CAS Number:
Molecular Weight:
452.57
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (857353P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857353P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCC/C=C\CCCCCCCC)N[C@@H](COP(O)([O-])=O)CO.[NH4+]

General description

VPC 31143 (R) functions as a lysophosphatidic acid (LPA)1/3 receptor agonist.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857353P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of lysophosphatidic acid (LPA) mediated stimulation of intestinal apical Cl-/OH- exchange
Singla A, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 298(2), G182-G189 (2009)
Hamed Horati et al.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society (2020-02-15)
Previously, we showed that abnormal levels of bioactive lipids in bronchoalveolar lavage fluid (BALF) from infants with cystic fibrosis (CF) correlated with early structural lung damage. To extend these studies, BALF bioactive lipid measurement by mass spectrometry and chest computed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service