Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

857134C

Avanti

18:1 Hemi BMP (S,R)

sn-(3-oleoyl-2-hydroxy)-glycerol-1-phospho-sn-3′-(1′,2′-dioleoyl)-glycerol (ammonium salt), chloroform

Synonym(s):

110734

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C60H114NO11P
CAS Number:
Molecular Weight:
1056.52
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (857134C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857134C

concentration

5 mg/mL (857134C-5mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OC[C@@]([H])(O)COC(CCCCCCC/C=C\CCCCCCCC)=O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

General description

18:1 Hemi BMP (S,R) is also called as acyl phosphatidylglycerol or hemi-bis (diacylglycero) phosphate. It has acyl chains at the sn-3 and sn-1′ positions, along with an additional acyl chain at position sn-2.[1]

Application

18:1 Hemi BMP (S,R) is suitable to study its interfacial physical properties using Langmuir monomolecular films.[1]

Biochem/physiol Actions

18:1 Hemi BMP (S,R) can acts as putative precursors of bis(monoacylglycero)phosphate (BMP).[1]

Packaging

5 mL Clear Glass Sealed Ampule (857134C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis (monoacylglycero) phosphate interfacial properties and lipolysis by pancreatic lipase-related protein 2, an enzyme present in THP-1 human monocytes
Record M, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1811(7-8), 419-430 (2011)
Qiang Cheng et al.
Nature nanotechnology, 15(4), 313-320 (2020-04-07)
CRISPR-Cas gene editing and messenger RNA-based protein replacement therapy hold tremendous potential to effectively treat disease-causing mutations with diverse cellular origin. However, it is currently impossible to rationally design nanoparticles that selectively target specific tissues. Here, we report a strategy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service