Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

855475C

Avanti

12:0 Lyso PC

1-lauroyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-dodecanoyl-sn-glycero-3-phosphocholine; PC(12:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H42NO7P
CAS Number:
Molecular Weight:
439.52
UNSPSC Code:
51191904
NACRES:
NA.25

assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855475C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855475C

concentration

10 mg/mL (855475C-25mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C20H42NO7P/c1-5-6-7-8-9-10-11-12-13-14-20(23)26-17-19(22)18-28-29(24,25)27-16-15-21(2,3)4/h19,22H,5-18H2,1-4H3/t19-/m1/s1

InChI key

BWKILASWCLJPBO-LJQANCHMSA-N

Application

12:0 Lyso PC may be used to reversibly block envelope glycoprotein (Env)-mediated fusion in various cell lines.[1] It may also be used in determining bubble local velocity profile.[2]

Packaging

5 mL Clear Glass Sealed Ampule (855475C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fusion stage of HIV-1 entry depends on virus-induced cell surface exposure of phosphatidylserine
Zaitseva E, et al.
Cell host & microbe, 22(1), 99-110 (2017)
Effect of products of PLA2 catalyzed hydrolysis of DLPC on motion of rising bubbles
Krzan M, et al.
Colloids and Surfaces. B, Biointerfaces, 128, 261-267 (2015)
Boris Fichtman et al.
Molecular biology of the cell, 21(23), 4197-4211 (2010-10-12)
Nuclear pore complexes (NPCs) are large proteinaceous channels embedded in double nuclear membranes, which carry out nucleocytoplasmic exchange. The mechanism of nuclear pore assembly involves a unique challenge, as it requires creation of a long-lived membrane-lined channel connecting the inner
Shi-Yuan Wen et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 69, 152690-152690 (2018-11-06)
Rhodioloside is a glucoside of tyrosol isolated from Rhodiola rosea. However, its regulating effect on hepatic dyslipidemia of atherogenic mice has rarely been studied. The specific aims of current study included to clarify lipidomic perturbation in liver tissues of apolipoprotein
Yutaka Ikeno et al.
The Journal of biological chemistry, 280(30), 28044-28052 (2005-06-02)
We previously demonstrated that secretory phospholipase A2 (sPLA2) and lysophosphatidylcholine (LPC) exhibit neurotrophin-like neuritogenic activity in the rat pheochromocytoma cell line PC12. In this study, we further analyzed the mechanism whereby sPLA2 displays neurite-inducing activity. Exogenously added mammalian group X

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service