Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

855375C

Avanti

10:0 Lyso PC

1-decanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-decanoyl-sn-glycero-3-phosphocholine; PC(10:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H38NO7P
CAS Number:
Molecular Weight:
411.47
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855375C-25mg)
pkg of 1 × 20 mL (855375C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855375C

concentration

10 mg/mL (855375C-25mg)
25 mg/mL (855375C-500mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C18H38NO7P/c1-5-6-7-8-9-10-11-12-18(21)24-15-17(20)16-26-27(22,23)25-14-13-19(2,3)4/h17,20H,5-16H2,1-4H3/t17-/m1/s1

InChI key

SECPDKKEUKDCPG-QGZVFWFLSA-N

Application

10:0 Lyso PC may be used as an internal standard to quantify lysophosphatidylcholine (LPC) species found in Trypanosoma cruzi samples.[1]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (855375C-500mg)
5 mL Clear Glass Sealed Ampule (855375C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and functional analysis of a platelet-activating lysophosphatidylcholine of Trypanosoma cruzi
Gazos-Lopes F, et al.
PLoS Neglected Tropical Diseases, 8(8) (2014)
Felipe Gazos-Lopes et al.
PLoS neglected tropical diseases, 8(8), e3077-e3077 (2014-08-08)
Trypanosoma cruzi is the causative agent of the life-threatening Chagas disease, in which increased platelet aggregation related to myocarditis is observed. Platelet-activating factor (PAF) is a potent intercellular lipid mediator and second messenger that exerts its activity through a PAF-specific
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service