Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

855276C

Avanti

09:0 Lyso PC

1-nonanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-nonanoyl-sn-glycero-3-phosphocholine; PC(9:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H36NO7P
CAS Number:
Molecular Weight:
397.44
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855276C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855276C

concentration

10 mg/mL (855276C-25mg)

shipped in

dry ice

Storage temp.

−20°C

General description

09:0 Lyso PC is a short-chain polar lysophosphatidylcholine (lysoPC).

Application

09:0 Lyso PC is suitable for use in thin-layer chromatography (TLC) analysis for lipid extraction.

Packaging

5 mL Clear Glass Sealed Ampule (855276C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preferential hydrolysis of truncated oxidized glycerophospholipids by lysosomal phospholipase A2
Abe A, et al.
Journal of Lipid Research, 58(2), 339-349 (2017)
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service