Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

855275C

Avanti

08:0 Lyso PC

1-octanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-octanoyl-sn-glycero-3-phosphocholine; PC(8:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H34NO7P
CAS Number:
Molecular Weight:
383.42
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855275C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855275C

concentration

10 mg/mL (855275C-25mg)

shipped in

dry ice

storage temp.

−20°C

application

08:0 Lyso PC may be used as a substrate for sphingomyelinase D-like protein (SicTox)in 31P nuclear magnetic resonance (31P NMR) assays.[1][2] It may also be used to study the self-aggregation properties of lysophosphatidylcholines (lysoPC′s).[3]

Packaging

5 mL Clear Glass Sealed Ampule (855275C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phospholipase D toxins of brown spider venom convert lysophosphatidylcholine and sphingomyelin to cyclic phosphates
Lajoie DM, et al.
PLoS ONE, 8(8) (2013)
Microstructural characterization of lysophosphatidylcholine micellar aggregates: the structural basis for their use as biomembrane mimics
Vitiello G, et al.
Journal of Colloid and Interface Science, 336(2), 827-833 (2009)
Variable substrate preference among phospholipase D toxins from Sicariid spiders
Lajoie DM, et al.
Test, 290(17), 10994-11007 (2015)
Phospholipase D toxins of brown spider venom convert lysophosphatidylcholine and sphingomyelin to cyclic phosphates
Lajoie DM, et al.
Testing, 8(8) (2013)
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service