Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850854C

Avanti

16:0 Dimethyl PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N,N-dimethyl, chloroform

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N,N-dimethyl; PDME; 110742; phosphatidyldimethylethanolamine;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H78NO8P
CAS Number:
Molecular Weight:
720.01
UNSPSC Code:
12352211
NACRES:
NA.25
Product 850854C is not currently sold in your country. Contact Technical Service

assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 2.5 mL (850854C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850854C

concentration

10 mg/mL (850854C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)[H])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H78NO8P/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-38(41)45-35-37(36-47-49(43,44)46-34-33-40(3)4)48-39(42)32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h37H,5-36H2,1-4H3,(H,43,44)

InChI key

SKWDCOTXHWCSGS-UHFFFAOYSA-N

General description

16:0 Dimethyl PE functions as an intermediate in the formation of the phosphatidylcholine (PC).[1]

Packaging

5 mL Clear Glass Sealed Ampule (850854C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rat brain phosphatidyl-N, N-dimethylethanolamine is rich in polyunsaturated fatty acids
Tacconi M, et al.
Journal of Neurochemistry, 45(3), 805-809 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service