Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850850C

Avanti

18:1 Monomethyl PE

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-methyl, chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine-N-methyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80NO8P
CAS Number:
Molecular Weight:
758.06
UNSPSC Code:
51191904

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850850C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850850C

concentration

10 mg/mL (850850C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)([H])[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C42H80NO8P/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43-3)51-42(45)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h18-21,40,43H,4-17,22-39H2,1-3H3,(H,46,47)/b20-18+,21-19+/t40-/m1/s1

InChI key

LPXFOQGBESUDAX-KFVQOHGZSA-N

General description

18:1 Monomethyl phosphoethanolamine (PE) is an alkyl phosphate derivative. Monomethyl PE is formed, when methyl transferases acts on PE. It might be implicated in the transduction of biochemical signals through the cell membrane.

Application

18:1 Monomethyl PE is suitable for the preparation of standard liposomes. It is also suitable to study its effect on the interaction of fructan with monomolecular lipid system.

Packaging

5 mL Clear Glass Sealed Ampule (850850C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane Fluidity (2012)
Grzegorz Pawlik et al.
The Journal of biological chemistry, 295(8), 2473-2482 (2020-01-15)
Phospholipid N-methyltransferases (PLMTs) synthesize phosphatidylcholine by methylating phosphatidylethanolamine using S-adenosylmethionine as a methyl donor. Eukaryotic PLMTs are integral membrane enzymes located in the endoplasmic reticulum (ER). Recently Opi3, a PLMT of the yeast Saccharomyces cerevisiae was proposed to perform in
Structural requirements of the fructan-lipid interaction
Vereyken IJ, et al.
Biophysical Journal, 84(5), 3147-3154 (2003)
Effects of lipids on the interaction of SecA with model membranes
Ahn T, et al.
Archives of Biochemistry and Biophysics, 395(1), 14-20 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service