Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

850581P

Avanti

LFA-C4

powder

Synonym(s):

Lithocholate-based Facial Amphiphile-C4

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C67H116O33
Molecular Weight:
1449.62
UNSPSC Code:
12352211

assay

99% (TLC)

form

powder

packaging

package of 1 × 200 mg
package of 1 × 25 mg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Lithocholate-based Facial Amphiphile-C4 (LFA-C4) is a facial detergent. It is water soluble upto 2.5%. LFA-C4 is more efficacious in stabilization of G-protein coupled receptors (GPCRs) than lauryl maltose neopentyl glycol (LMNG). It is also more effective in receptor stability than n-dodecyl-β-d-maltoside (DDM) and lithocholate-based bola-amphiphile-C0 (LBA-C0) for leucine transporter (LeuT) and bacteriorhodopsin (Br).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (850581P-25MG)
20 mL Clear Glass Screw Cap Vial (850581P-200MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Engineered Lithocholate-Based Facial Amphiphile Stabilizes Membrane Proteins: Assessing the Impact of Detergent Customizability on Protein Stability
Das M, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 24(39), 9860-9868 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service