Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850474C

Avanti

18:1-14:0 PC

1-oleoyl-2-myristoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1--(9Z-octadecenoyl)-2-tetradecanoyl-sn-glycero-3-phosphocholine; OMPC; PC(18:1(9Z)/14:0)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H78NO8P
CAS Number:
Molecular Weight:
732.02
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850474C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850474C

concentration

10 mg/mL (850474C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is an important component of the mucosal layer of the colon[1] and is the most common phospholipid in mammalian membranes. It is a strong bilayer-forming lipid.[2]

Application

18:1-14:0 PC has been used as a phospholipid standard to extract phospholipid fatty acids (PLFAs).[3]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[1] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850474C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of amino acid indices for assessing organic matter quality and microbial abundance in deep-sea Antarctic sediments of IODP Expedition 318
Carr SA, et al.
Marine Chemistry, 186, 72-82 (2016)
The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service