Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850344C

Avanti

14:0-04:0(biphenyl) PC (TBBPC)

1-myristoyl-2-[4-(4-biphenyl)butanoyl]-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

TBBPC

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H60NO8P
CAS Number:
Molecular Weight:
689.86
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850344C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850344C

concentration

10 mg/mL (850344C-10mg)

shipped in

dry ice

storage temp.

−20°C

General description

14:0-04:0(biphenyl) PC (TBBPC/1-myristoyl-2-[4-(4-biphenyl)butanoyl]-sn-glycero-3-phosphocholine) is a long-chain biphenyl phospholipid.[1]

Application

14:0-04:0(biphenyl) PC (TBBPC/1-myristoyl-2-[4-(4-biphenyl)butanoyl]-sn-glycero-3-phosphocholine) may be used in bicelle preparations.[2]

Biochem/physiol Actions

14:0-04:0(biphenyl) PC (TBBPC/1-myristoyl-2-[4-(4-biphenyl)butanoyl]-sn-glycero-3-phosphocholine) bicelles may be used for metal binding membrane protein studies.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850344C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna A De Angelis et al.
Biophysical journal, 101(5), 1086-1094 (2011-09-06)
The amphipathic antimicrobial peptide piscidin 1 was studied in magnetically aligned phospholipid bilayers by oriented-sample solid-state NMR spectroscopy. (31)P NMR and double-resonance (1)H/(15)N NMR experiments performed between 25 °C and 61 °C enabled the lipid headgroups as well as the
Cécile Loudet-Courreges et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9122-9130 (2011-06-15)
Nanometric bilayer-based self-assembled micelles commonly named as bicelles, formed with a mixture of long and short chains phosphatidylcholine lipids (PC), are known to orient spontaneously in a magnetic field. This field-induced orientational order strongly depends on the molecular structure of
Sang Ho Park et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 193(1), 133-138 (2008-05-22)
Bicelles composed of the long-chain biphenyl phospholipid TBBPC (1-tetradecanoyl-2-(4-(4-biphenyl)butanoyl)-sn-glycero-3-PC) and the short-chain phospholipid DHPC align with their bilayer normals parallel to the direction of the magnetic field. In contrast, in typical bicelles the long-chain phospholipid is DMPC or DPPC, and
Matthieu Fillion et al.
Biochimica et biophysica acta, 1838(9), 2173-2179 (2014-02-11)
We have investigated in the present study the effect of both non-selective and selective cationic 14-mer peptides on the lipid orientation of DMPC bilayers by (31)P solid-state nuclear magnetic resonance (NMR) spectroscopy. Depending on the position of substitution, these peptides
Anna Diller et al.
Biochimie, 91(6), 744-751 (2009-03-03)
Major biological processes occur at the biological membrane. One of the great challenges is to understand the function of chemical or biological molecules inside the membrane; as well of those involved in membrane trafficking. This requires obtaining a complete picture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service