Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850330C

Avanti

11:0 PC

1,2-diundecanoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

PC(11:0/11:0)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H60NO8P
CAS Number:
Molecular Weight:
593.77
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850330C-25mg)
pkg of 1 × 20 mL (850330C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850330C

concentration

10 mg/mL (850330C-25mg)
25 mg/mL (850330C-500mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C30H60NO8P/c1-6-8-10-12-14-16-18-20-22-29(32)36-26-28(27-38-40(34,35)37-25-24-31(3,4)5)39-30(33)23-21-19-17-15-13-11-9-7-2/h28H,6-27H2,1-5H3

InChI key

XCCDCYFQYUARAY-UHFFFAOYSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes.[1] It is also an important component of the mucosal layer of the colon.[2]
Phosphatidylcholine (PC) is the chief phospholipid present in the mammalian tissues. It accounts to 50% of total phospholipid content of the cell. PC has one head comprising choline and phosphate; two tails of fatty acids.[3] Generally, PC from mammalian tissues contains saturated fatty acid in the C1 position and unsaturated fatty acyl in the C2 position of the glycerol.[4]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

11:0 PC (1,2-diundecanoyl-sn-glycero-3-phospho choline) has been used:
  • to supplement with bacteria to feed L4 larvae for Caenorhabditis elegans fat content measurement[5]
  • in the preparation of liposomes[6][7]
  • as an internal standard during lipid extraction and subsequent liquid chromatography-mass spectrometry (LC-MS) analysis[8]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[2] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[9]
Phosphatidylcholine (PC) synthesis is stimulated by certain hormones, growth factors and phorbol esters by the action of phosphocholine cytidyltransferase. Hydrolysis of PC plays a key role in cell signaling.[10]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850330C-500mg)
5 mL Clear Glass Sealed Ampule (850330C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Liver,Kidney, Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingolipid metabolism correlates with cerebrospinal fluid Beta amyloid levels in Alzheimer?s disease
Fonteh AN, et al.
PLoS ONE, 10(5), e0125597-e0125597 (2015)
The Membranes of Cells (2016)
Skp is a multivalent chaperone of outer-membrane proteins
Schiffrin B, et al.
Nature Structural and Molecular Biology, 23(9), 786-786 (2016)
The regulation and cellular functions of phosphatidylcholine hydrolysis.
Billah MM and Anthes JC
The Biochemical Journal, 269(2), 281-281 (1990)
Integrative Medicine - E-Book (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service