Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850312C

Avanti

DOCPe

Avanti Research - A Croda Brand 850312C

Synonym(s):

2-((2,3-bis(oleoyloxy)propyl)dimethylammonio)ethyl ethyl phosphate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H86NO8P
CAS Number:
Molecular Weight:
800.14
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850312C-25mg)
pkg of 2 × 4 mL (850312C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850312C

concentration

10 mg/mL (850312C-25mg)
25 mg/mL (850312C-200mg)

shipped in

dry ice

storage temp.

−20°C

General description

2-((2,3-bis(oleoyloxy)propyl)dimethylammonio)ethyl ethylphosphate (DOCPe) is a neutrally charged lipid.[1]
Inverse-phosphocholine lipids, a remix of a common phospholipid, contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Therefore iPC lipids afford a unique opportunity to investigate the biophysical and bioactivity-related ramifications of a charge inversion at the bilayer surface.

Application

DOCPe is suitable for liposomes preparation.[1] It is also suitable to use as membrane to examine the impact of the phospholipid head on peptide-membrane interaction.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850312C-200mg)
5 mL Clear Glass Sealed Ampule (850312C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Liver,Kidney, Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partition of Positively and Negatively Charged Tryptophan Ions in Membranes with Inverted Phospholipid Heads: Simulations and Experiments
Cardenas AE, et al.
The Journal of Physical Chemistry B, 123(15), 3272-3281 (2019)
Charge and Coordination Directed Liposome Fusion onto SiO2 and TiO2 Nanoparticles
Wang X, et al.
Langmuir (2018)
Yibo Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 34(32), 9337-9348 (2018-03-13)
Phospholipids are a major component of the cell membrane. In most natural phospholipids, the phosphate acts as a bridge, connecting the other portion of the polar headgroup with the hydrophobic tails. Such bridging phosphate is chemically quite inert. Synthetic lipids
Emily K Perttu et al.
Journal of the American Chemical Society, 134(10), 4485-4488 (2012-03-01)
Zwitterionic inverse-phosphocholine (iPC) lipids contain headgroups with an inverted charge orientation relative to phosphocholine (PC) lipids. The iPC lipid headgroup has a quaternary amine adjacent to the bilayer interface and a phosphate that extends into the aqueous phase. Neutral iPC
Feng Wang et al.
Journal of the American Chemical Society, 137(36), 11736-11742 (2015-08-25)
Zwitterionic phosphocholine (PC) lipids are highly biocompatible, representing a major component of the cell membrane. A simple mixing of PC liposomes and silica (SiO2) surface results in liposome fusion with the surface and formation of supported lipid bilayers. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service