850304P
Avanti
05:0 PC
1,2-dipentanoyl-sn-glycero-3-phosphocholine, powder
Synonym(s):
1,2-divaleryl-sn-glycero-3-phosphocholine; PC(5:0/5:0)
Select a Size
Select a Size
About This Item
Recommended Products
assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 25 mg (850304P-25mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 850304P
shipped in
dry ice
storage temp.
−20°C
SMILES string
[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCC)=O)COC(CCCC)=O)=O
General description
Application
- as a component in the preparation of endophilin Bin−Amphiphysin−Rvs domain (EndoBAR) deforms membranes[2]
- in binary mixtures to study the effects of lipid chain length and unsaturation on bicelles stability[3]
- in the preparation of homo and heterooligomers for ESi MSMS studies[4]
Biochem/physiol Actions
Packaging
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service