Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

840036C

Avanti

10:0 PS

1,2-didecanoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Synonym(s):

PS(10:0/10:0)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H49NO10PNa
CAS Number:
Molecular Weight:
589.63
UNSPSC Code:
51191904
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (840036C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840036C

concentration

10 mg/mL (840036C-10mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCC)=O)COC(CCCCCCCCC)=O.[Na+]

InChI

1S/C26H50NO10P.Na/c1-3-5-7-9-11-13-15-17-24(28)34-19-22(20-35-38(32,33)36-21-23(27)26(30)31)37-25(29)18-16-14-12-10-8-6-4-2;/h22-23H,3-21,27H2,1-2H3,(H,30,31)(H,32,33);/q;+1/p-1/t22-,23+;/m1./s1

InChI key

MNHRZRUSKRPBDA-RFPXDPOKSA-M

General description

Glycerophosphoserines (PS) interacts with spectrin and maintains red blood cell (RBC) membrane asymmetry.[1]

Application

1,2-didecanoyl-sn-glycero-3-phospho-L-serine has been used for the preparation of sucrose-loaded liposomes.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840036C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding of PLD2-generated phosphatidic acid to KIF5B promotes MT1-MMP surface trafficking and lung metastasis of mouse breast cancer cells
Wang Z, et al.
Developmental Cell, 43(2), 186-197 (2017)
Phospholipidomics reveals differences in glycerophosphoserine profiles of hypothermically stored red blood cells and microvesicles
Bicalho B, et al.
Biochimica et Biophysica Acta - Biomembranes, 1828(2), 317-326 (2013)
Emily R Schenk et al.
Journal of mass spectrometry : JMS, 50(1), 88-94 (2015-01-21)
Bacterial resistance to antibiotics has become an increasing threat, requiring not only the development of new targets in drug discovery, but more importantly, a better understanding of cellular response. In the current study, three closely related Escherichia coli strains, a
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service