Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810608P

Avanti

18:1 Tempo PC

Avanti Research - A Croda Brand 810608P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phospho(tempo)choline

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C52H98N2O9P
CAS Number:
Molecular Weight:
926.32
UNSPSC Code:
12352211

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810608P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810608P

lipid type

ESR probes

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C1CC(C)(C)N([O])C(C)(C)C1)C)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

General description

18:1 Tempo PC is a stable isotope and acts as an erythrocyte sedimentation rate (ESR) probe.

Packaging

5 mL Clear Glass Sealed Ampule (810608P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhagyashree D Rao et al.
Chemistry and physics of lipids, 226, 104849-104849 (2019-12-15)
pH (low) insertion peptide (pHLIP) is a polypeptide from the third transmembrane helix of bacteriorhodopsin. The pH-dependent membrane insertion of pHLIP has been conveniently exploited for translocation of cargo molecules and as a novel imaging agent in cancer biology due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service