Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810607P

Avanti

18:1 TEMPO Lyso PC

Avanti Research - A Croda Brand 810607P, powder

Synonym(s):

1-oleoyl-2-hydroxy-sn-glycero-3-phospho(tempo)choline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H66N2O8P
CAS Number:
Molecular Weight:
661.87
UNSPSC Code:
41141825
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810607P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810607P

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidylcholine (LPC) is one of the important constituent of oxidized low-density lipoprotein (Ox-LDL). 1-oleoyl-2-hydroxy-sn-glycero-3-phospho(tempo)choline (18:1 TEMPO Lyso PC), a lipoxygenase substrate analog is a lysooleoyllecithin which carries a spin label instead of a choline methyl. It is a paramagnetic lysolipid.
TEMPO and DOXYL spin probes penetrate liposome membranes. This may reveal new components with increased ordering of the lipids associated with peptides or proteins.

Application

1-oleoyl-2-hydroxy-sn-glycero-3-phospho(tempo)choline (18:1 TEMPO Lyso PC) may be used as a spin-labeled lysolecithin (LOPTC) in micelles to study the nature of lipoxygenase interaction by electron paramagnetic resonance spectroscopy.

Biochem/physiol Actions

Lysophosphatidylcholine (LPC) is known to play a vital role in the atherogenic activity of oxidized low-density lipoprotein (Ox-LDL). 1-oleoyl-2-hydroxy-sn-glycero-3-phospho(tempo)choline (18:1 TEMPO Lyso PC) can form micelles in solution.

Packaging

5 mL Clear Glass Sealed Ampule (810607P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takayuki Matsumoto et al.
Current medicinal chemistry, 14(30), 3209-3220 (2008-01-29)
Lysophosphatidylcholine (LPC) is a bioactive proinflammatory lipid generated by pathological activities. LPC is also a major phospholipid component of oxidized low-density lipoprotein (Ox-LDL) and is implicated as a critical factor in the atherogenic activity of Ox-LDL. LPC is believed to
Betty J Gaffney et al.
Biophysical journal, 103(10), 2134-2144 (2012-12-04)
Lipoxygenase enzymes initiate diverse signaling pathways by specifically directing oxygen to different carbons of arachidonate and other polyunsaturated acyl chains, but structural origins of this specificity have remained unclear. We therefore determined the nature of the lipoxygenase interaction with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service