Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810605P

Avanti

16:0-14 Doxyl PC

Avanti Research - A Croda Brand 810605P, powder

Synonym(s):

1-palmitoyl-2-stearoyl-(14-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810605P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810605P

shipped in

dry ice

storage temp.

−20°C

General description

16:0-14 Doxyl Phosphatidylcholine (PC) is a doxyl probe with a nitroxyl ring attached at carbon 14 on the chain of 1-palmitoyl-2-stearoyl-(14-doxyl)-sn-glycero-3-phosphocholine (PSPC).[1][2]
Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins and as fluorescent quenchers in lipid bilayer structural studies.[3][4]

application

16:0-14 Doxyl PC (1-palmitoyl-2-stearoyl-(14-doxyl)-sn-glycero-3-phosphocholine) has been used:
  • in the labeling of small unilamellar vesicles for electron spin resonance (ESR) spectra[5]
  • as spin label for labeling at different depths of 1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine (PSPC) multilamellar vesicles (MLVs) for nuclear magnetic resonance (NMR) studies[1]
  • as paramagnetic spin labels for 1H-15N heteronuclear single quantum coherence (HSQC) spectra[2]

Biochem/physiol Actions

16:0-14 Doxyl Phosphatidylcholine (PC) is used as a label to probe various membrane depths from the hydrocarbon-water interface.[5] It quenches the fluorescence of 7-nitrobenz-2-oxa-1,3-diazole (NBD), a fluorescent label in the lipid bilayer.[6] There is a progressive increase in the depth and degree of mobility of the probe as the doxyl group substitution proceeds towards the end of the lipid chain,[6][7] and flexibility also increases.[7]

Packaging

5 mL Clear Glass Sealed Ampule (810605P-1mg)

Preparation Note

Product use: To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies.[8] For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM [3], Additional supplemental information.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-state NMR paramagnetic relaxation enhancement immersion depth studies in phospholipid bilayers
Chu S, et al.
Journal of Magnetic Resonance, 207(1), 89-94 (2010)
Molecular mechanism of membrane docking by the Vam7p PX domain
Lee SA, et al.
The Journal of Biological Chemistry, 281(48), 37091-37101 (2006)
Molecular dynamics simulations of depth distribution of spin-labeled phospholipids within lipid bilayer
Kyrychenko A and Ladokhin AS
The Journal of Physical Chemistry B, 117(19), 5875-5885 (2013)
Calibration of distribution analysis of the depth of membrane penetration using simulations and depth-dependent fluorescence quenching
Kyrychenko A, et al.
The Journal of Membrane Biology, 248(3), 583-594 (2015)
Binding of alpha-synuclein affects the lipid packing in bilayers of small vesicles
Kamp, et al.
Test, 281(14), 9251-9259 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service