Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810605C

Avanti

16:0-14 Doxyl PC

Avanti Research - A Croda Brand 810605C

Synonym(s):

1-palmitoyl-2-stearoyl-(14-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810605C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810605C

concentration

1 mg/mL (810605C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins and as fluorescent quenchers in lipid bilayer structural studies.[1][2]

application

16:0-14 Doxyl PC has been used:
  • as a spin label for electron paramagnetic resonance (EPR) studies[3]
  • in labeling of small unilamellar vesicles (SUV) for electron spin resonance (ESR) studies[4]
  • in polarity profile studies of phospholipid membranes by high-frequency ESR[5]

Biochem/physiol Actions

16:0-14 Doxyl phosphatidylcholine (PC) is used in the labeling of small unilamellar vesicles (SUVs) to probe the membrane depth from the hydrocarbon-water interface.[4] n-Doxyl PC is incorporated in membranes for depth quenching measurements. It is used in fluorescence based vesicle quenching studies.[6]

Packaging

5 mL Clear Glass Sealed Ampule (810605C-1mg)

Preparation Note

Product use: To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies.[7] For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM [1], Additional supplemental information.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spin-label saturation-recovery EPR at W-band: applications to eye lens lipid membranes
Mainali L, et al.
Journal of Magnetic Resonance, 212(1), 86-94 (2011)
Conformational distributions and hydrogen bonding in gel and frozen lipid bilayers: a high frequency spin-label ESR study
Dzikovski B, et al.
The Journal of Physical Chemistry B, 116(23), 6694-6706 (2012)
Basis for selectivity of cationic antimicrobial peptides for bacterial versus mammalian membranes
Glukhov E, et al.
The Journal of Biological Chemistry, 280(40), 33960-33967 (2005)
Binding of alpha-synuclein affects the lipid packing in bilayers of small vesicles
Kamp, et al.
Test, 281(14), 9251-9259 (2006)
Diego E Sastre et al.
The Journal of biological chemistry, 295(7), 2136-2147 (2019-12-05)
PlsX plays a central role in the coordination of fatty acid and phospholipid biosynthesis in Gram-positive bacteria. PlsX is a peripheral membrane acyltransferase that catalyzes the conversion of acyl-ACP to acyl-phosphate, which is in turn utilized by the polytopic membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service