Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810602C

Avanti

16:0-7 Doxyl PC

Avanti Research - A Croda Brand 810602C

Synonym(s):

1-palmitoyl-2-stearoyl-(7-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
UNSPSC Code:
41141825
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810602C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810602C

concentration

1 mg/mL (810602C-1mg)

lipid type

ESR probes

shipped in

dry ice

storage temp.

−20°C

General description

16:0-7 Doxyl PC is a nitroxide labeled phospholipid. It is an uncharged hydrophobic quencher. It is restricted to hydrophobic core of the membrane.[1]
Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins and as fluorescent quenchers in lipid bilayer structural studies. [2][3]
The excretory and secretory products of helminths has a small hapten-like portion called phosphorylcholine (PC).[4] Several prokaryotic and eukaryotic pathogens has PC as its structural constituent.[5]

Application

16:0-7 Doxyl PC (1-palmitoyl-2-stearoyl-(7-doxyl)-sn-glycero-3-phosphocholine)(7-NO-PC) has been used:
  • in liposome preparation[1]
  • as a spin-labeled probe for quenching analysis[6]
  • in the preparation of spin-labeled lipid vesicles for electron paramagnetic resonance (EPR) spectroscopy[7]
  • in the preparation of liposomes for fluorescence quenching experiments[8]

Biochem/physiol Actions

Phosphorylcholine (PC) possesses immunomodulatory properties.[5]

Packaging

5 mL Clear Glass Sealed Ampule (810602C-1mg)

Preparation Note

Product use: To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies.[9] For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM.[3] , Supplemental information

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational changes in BID, a pro-apoptotic BCL-2 family member, upon membrane binding a SITE-DIRECTED SPIN LABELING STUDY
Oh KJ, et al.
The Journal of Biological Chemistry, 280(1), 753-767 (2005)
Dynamic behavior of fatty acid spin labels within a binding site of soybean lipoxygenase-1
Wu F, et. al.
Biochemistry, 45, 12510-12518 (2006)
The mechanism of membrane insertion for a cholesterol-dependent cytolysin: a novel paradigm for pore-forming toxins
Shatursky O, et al.
Cell, 99(3), 293-299 (1999)
Host defenses to helminths
Clinical Immunology of the Dog and Cat, 367-377 (2013)
Viewing membrane-bound molecular umbrellas by parallax analyses.
Kondo M, et. al.
Journal of the American Chemical Society, 130, 13771-13777 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service