Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810590O

Avanti

10-Doxyl Nonadecane

Avanti Research - A Croda Brand 810590O

Synonym(s):

10-doxyl nonadecane

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H46NO2
CAS Number:
Molecular Weight:
368.62
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mg (810590O-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810590O

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C23H46NO2/c1-5-7-9-11-13-15-17-19-23(24(25)22(3,4)21-26-23)20-18-16-14-12-10-8-6-2/h5-21H2,1-4H3

InChI key

LMFAKDOCBCXKIK-UHFFFAOYSA-N

General description

Quenching of tryptophan (Trp) by 10-docylnonadecane can be used to measure Trp depth in the lipid membrane.

Packaging

5 mL Clear Glass Sealed Ampule (810590O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W R Perkins et al.
International journal of pharmaceutics, 200(1), 27-39 (2000-06-14)
The development of stable spherical lipid-coated drug particles that are termed 'lipocores' is reported here. Unlike conventional lipid-based particles (i.e. liposomes, emulsions, micelles), these particles are comprised solely of a core of a poorly water soluble drug surrounded by polyethyleneglycol
Devaki A Kelkar et al.
Biochemical and biophysical research communications, 343(2), 483-488 (2006-03-21)
The linear peptide gramicidin forms prototypical ion channels specific for monovalent cations and has been extensively used to study the organization, dynamics, and function of membrane-spanning channels. We have analyzed the localization of the functionally important tryptophan residues of the
David J Shirley et al.
Peptide science (Hoboken, N.J.), 110(4) (2019-01-15)
Amphiphilic alpha-helices are common motifs used in numerous biological systems including membrane channels/pores and antimicrobial peptides (AMPs), and binding proteins, and a variety of synthetic biomaterials. Previously, an amphiphilic peptide with lysine-containing motifs was shown to reversibly bind the anionic
Gregory A Caputo et al.
Biochemistry, 42(11), 3265-3274 (2003-03-19)
A novel fluorescence method for determining the depth of Trp residues in membrane-inserted polypeptides is introduced. Quenching of Trp by acrylamide and 10-doxylnonadecane (10-DN) was used to measure Trp depth. Transmembrane helices with Trp residues at varying positions (and thus
Abhishek Chatterjee et al.
The Journal of biological chemistry, 290(42), 25579-25594 (2015-09-02)
Enterohemorrhagic Escherichia coli is a causative agent of gastrointestinal and diarrheal diseases. Pathogenesis associated with enterohemorrhagic E. coli involves direct delivery of virulence factors from the bacteria into epithelial cell cytosol via a syringe-like organelle known as the type III

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service