Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810509C

Avanti

18:1 Dabsyl PE

Avanti Research - A Croda Brand 810509C

Synonym(s):

1,2-dioleyl-sn-glycero-3-phosphoethanolamine-N-(dabsyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C55H94N5O10PS
CAS Number:
Molecular Weight:
1048.40
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810509C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810509C

concentration

5 mg/mL (810509C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoethanolamine (PHOS)/ (PE) is a precursor to phosphatidylcholine and phosphatidylethanolamine.[1]

Biochem/physiol Actions

Phosphatidylethanolamine controls membrane fluidity in eukaryotic cells.[2] Phosphoethanolamine (PHOS) has the ability to repress tumour growth both in vitro and in vivo.[1] It possesses anticarcinogenic effects. It can also prevent the mRNA expression of cyclin D1 and VEGFR1 (vascular endothelial growth factor receptor 1).[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810509C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic phosphoethanolamine: the state of the art of scientific production
Anastacio L de B, et al.
Brazilian Journal of Pharmaceutical Sciences , 53(4), 3658-3667 (2017)
Phosphoethanolamine and the danger of unproven drugs
Ponde N, et al.
Ecancermedicalscience, 10(7), 3658-3667 (2016)
Phosphatidylethanolamine is a key regulator of membrane fluidity in eukaryotic cells
Dawaliby R, et al.
The Journal of Biological Chemistry, 291(7), 3658-3667 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service