Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810406M

Avanti

TopFluor-POVPE (DMA)

Avanti Research - A Croda Brand 810406M, methylene chloride solution

Synonym(s):

1-palmitoyl-2-(5,5-dimethoxyvaleroyl)-sn-glycero-3-phosphoethanolamine-N- [4-(dipyrrometheneboron difluoride)butanoyl] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H72BF2N4O10P
Molecular Weight:
884.83
UNSPSC Code:
12352202

assay

>99% (TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (810406M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810406M

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (810406M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service