Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810334C

Avanti

16:0 Pyrene PE

Avanti Research - A Croda Brand 810334C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(1-pyrenesulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C53H85 N2O10PS
Molecular Weight:
973.29
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810334C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810334C

concentration

1 mg/mL (810334C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Pyrene is made up of four fused benzol rings and is an apolar molecule.[1] Phosphatidylethanolamine (PE), a phospholipid is found abundantly in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells. It is especially abundant in the inner mitochondrial membrane.[2]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) can serve as a lipid chaperone that supports in the folding of membrane proteins. It plays a key role in the activity of respiratory complexes. PE also participates in the initiation of autophagy.[2] Pyrene-labeled PE may be used for liposome preparations.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810334C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

beta-Carotene-Lipid Interactions in Liposomes with Different Lipid Composition
Advances in Planar Lipid Bilayers and Liposomes, 14, 293-307 (2011)
T Ahn et al.
The Journal of biological chemistry, 273(34), 21692-21698 (1998-08-15)
The effect of nonlamellar-prone lipids, diacylglycerol (DG) and phosphatidylethanolamine (PE), on the ATPase activity of SecA was examined. When Escherichia coli PE of the standard vesicles composed of 60 mol% of this lipid and 40 mol% of dioleoylphosphatidylglycerol (DOPG) is
Dhaval Patel et al.
Oxidative medicine and cellular longevity, 2017, 4829180-4829180 (2017-08-09)
Phosphatidylethanolamine (PE) is the second most abundant phospholipid in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells; it is enriched in the inner leaflet of membranes, and it is especially abundant in the inner mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service