Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810306M

Avanti

PyrOVPC (DMA)

Avanti Research - A Croda Brand 810306M, methylene chloride solution

Synonym(s):

1-(10-pyrenedecanoyl)-2-(5,5-dimethoxyvaleroyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C41H58NO10P
CAS Number:
Molecular Weight:
755.87
UNSPSC Code:
12352202

assay

>99% (TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (810306M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810306M

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (810306M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service