Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810296C

Avanti

C11 TopFluor MG

Avanti Research - A Croda Brand

Synonym(s):

1-[11-(dipyrrometheneboron difluoride)undecanoyl]-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H41 BF2N2O4
CAS Number:
Molecular Weight:
506.43
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810296C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (810296C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Lipid droplets (LDs) are organelles that store lipids as reservoirs of metabolic energy and membrane lipid precursors, and they have been identified in the nucleus of some cells. LDs can be found in most eukaryotic cells. Most prominent are LDs in adipocytes, which make up most of the cellular volume, but other metabolically active cell types, such as liver or muscle cells, also have abundant LDs. LDs are unusual organelles in that they are bound by a monolayer of surface phospholipids, into which specific proteins are embedded, such as perilipins and metabolic enzymes. The cell biology of LDs as cellular organelles is only beginning to be unraveled. Avanti now offers a series of fluorescently-labeled mono-, di-, and triglycerides, which are useful for studying these important cellular organelles.

Packaging

5 mL Clear Glass Sealed Ampule (810296C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert V Farese et al.
Cell, 139(5), 855-860 (2009-12-01)
Long underappreciated as important cellular organelles, lipid droplets are finally being recognized as dynamic structures with a complex and interesting biology. In light of this newfound respect, we discuss emerging views on lipid droplet biology and speculate on the major
Nora Kory et al.
Trends in cell biology, 26(7), 535-546 (2016-03-21)
How proteins specifically localize to the phospholipid monolayer surface of lipid droplets (LDs) is being unraveled. We review here the major known pathways of protein targeting to LDs and suggest a classification framework based on the localization origin for the
Yuki Ohsaki et al.
The Journal of cell biology, 212(1), 29-38 (2016-01-06)
Lipid droplets (LDs) in the nucleus of hepatocyte-derived cell lines were found to be associated with premyelocytic leukemia (PML) nuclear bodies (NBs) and type I nucleoplasmic reticulum (NR) or the extension of the inner nuclear membrane. Knockdown of PML isoform
Nora Kory et al.
Developmental cell, 34(3), 351-363 (2015-07-28)
Lipid droplets (LDs) are lipid storage organelles that grow or shrink, depending on the availability of metabolic energy. Proteins recruited to LDs mediate many metabolic functions, including phosphatidylcholine and triglyceride synthesis. How the LD protein composition is tuned to the supply
Robert V Farese et al.
The Journal of cell biology, 212(1), 7-8 (2016-01-06)
Lipid droplets (LDs) are sometimes found in the nucleus of some cells. In this issue, Ohsaki et al. (2016. J. Cell Biol. http://dx.doi.org/10.1083/jcb.201507122) show that the nuclear membrane, promyelocytic leukemia bodies, and the protein PML-II play a role in nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service