Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810289P

Avanti

16:0-C4 TopFluor DNP Cap PE

1-palmitoyl-2-[4-(dipyrrometheneboron difluoride)butanoyl]-sn-glycero-3-phosphoethanolamine-N-((6-(2,4-dinitrophenyl)amino)hexanoyl) (ammonium salt), powder

Synonym(s):

PED6

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H79 BF2N7O13P
CAS Number:
Molecular Weight:
1065.98
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810289P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810289P

shipped in

dry ice

storage temp.

−20°C

General description

16:0-C4 TopFluor DNP Cap PE/PED6 is a light sensitive and non-hygroscopic phospholipid.[1] TopFluor, a fluorescent molecule carries dipyrromethene boron difluoride moiety. It is linked to several lipid molecules by means of covalent attachment.[2]

Biochem/physiol Actions

Intestinal lipid processing in zebrafish mutants can be assayed with the help of 16:0-C4 TopFluor DNP Cap PEA/PED6.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810289P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo imaging of zebrafish digestive organ function using multiple quenched fluorescent reporters
Hama K, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 296(2), G445-G453 (2009)
Investigation of the biophysical properties of a fluorescently modified ceramide-1-phosphate
Shirey CM, et al.
Chemistry and Physics of Lipids, 200, 32-32 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service