Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810289C

Avanti

16:0-C4 TopFluor DNP Cap PE

1-palmitoyl-2-[4-(dipyrrometheneboron difluoride)butanoyl]-sn-glycero-3-phosphoethanolamine-N-((6-(2,4-dinitrophenyl)amino)hexanoyl) (ammonium salt), chloroform

Synonym(s):

PED6

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H79 BF2N7O13P
CAS Number:
Molecular Weight:
1065.98
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810289C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810289C

concentration

1 mg/mL (810289C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

16:0-C4 TopFluor DNP Cap PE is a haptenated lipid.[1] TopFluor, a fluorescent molecule carries dipyrromethene boron difluoride moiety. It is linked to several lipid molecules by means of covalent attachment.[2]

Biochem/physiol Actions

Intestinal lipid processing in zebrafish mutants can be assayed with the help of 16:0-C4 TopFluor DNP Cap PEA/PED6.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810289C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interactions of antigen-sensitized liposomes with immobilized antibody: a homogeneous solid-phase immunoliposome assay.
Ho RJ and Huang L
Journal of immunology (Baltimore, Md. : 1950), 134(6), 4035-4040 (1985)
In vivo imaging of zebrafish digestive organ function using multiple quenched fluorescent reporters
Hama K, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 296(2), G445-G453 (2009)
Investigation of the biophysical properties of a fluorescently modified ceramide-1-phosphate
Shirey CM, et al.
Chemistry and Physics of Lipids, 200, 32-32 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service