Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810233P

Avanti

C12-NBD L-threo-Sphingosine

Avanti Research - A Croda Brand 810233P, powder

Synonym(s):

N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-L-threo-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H61N5O6
CAS Number:
Molecular Weight:
659.90
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (810233P-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810233P

shipped in

dry ice

storage temp.

−20°C

General description

C12-NBD L-threo-Sphingosine is a fluorescent derivative of L-threo-sphingosine.[1] Sphingosine, an amino alcohol is a part of structurally diverse group of lipids called sphingolipids.[2][3] L-threo-sphingosine is an isomer of sphingosine that can be distinguished by CD spectroscopy.[4]

Biochem/physiol Actions

Sphingosine is a product of de-acylation of ceramide.[2] Many studies have indicated that sphingosine mediates processes like senescence, apoptosis and differentiation by functioning as a tumor-suppressor lipid.[5] The addition of sphingosine to cell culture media is known to activate stress-activated protein kinases (SAPKs) such as Jun amino-terminal kinases (JNK) and p38, and inhibit the activity of the mitogen activated protein kinases (MAPKs), extracellular signal-regulated kinases (Erk-1 and Erk-2).[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810233P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Carbonate: A Versatile Material, 58(8), 462-466 (2016)
Medical Applications of Mass Spectrometry (2008)
Medical Applications of Mass Spectrometry (2008)
Joanna Woodcock
IUBMB life, 58(8), 462-466 (2006-08-19)
The sphingomyelin pathway is activated in response to many apoptotic stimuli leading to the accumulation of breakdown products ceramide and sphingosine. Ceramide has received much attention as an apoptotic second messenger whereas sphingosine has largely been overlooked as a mediator
Raghavendra Pralhada Rao et al.
Journal of lipids, 2013, 178910-178910 (2013-08-29)
Sphingolipids, a family of membrane lipids, are bioactive molecules that participate in diverse functions controlling fundamental cellular processes such as cell division, differentiation, and cell death. Given that most of these cellular processes form the basis for several pathologies, it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service