Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810227C

Avanti

C12-NBD Lactosyl Ceramide

Avanti Research - A Croda Brand 810227C

Synonym(s):

N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-D-lactosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C48H81N5O16
CAS Number:
Molecular Weight:
984.18
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810227C-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810227C

concentration

0.05 mg/mL (810227C-50ug)

shipped in

dry ice

storage temp.

−20°C

General description

C12-NBD Lactosyl ceramide is a fluorescent derivative of the naturally occurring compound lactosyl ceramide, which is present on neutrophils and macrophages.[1][2]

Biochem/physiol Actions

Lactosyl ceramide plays a key role in signaling cascades leading to phenotypic changes such as adhesion, migration, cell proliferation, and angiogenesis. In vertebrates, lactosyl ceramide is a major precursor for biosynthesis of glycosphingolipids involved in various vital cellular processes.[2][3][4][5] Lactosyl ceramide is also known to be crucial for mediating osteoclastogenesis.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (810227C-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iwamoto et al.
The Journal of biological chemistry, 276(49), 46031-46038 (2001-10-11)
Glycosphingolipids and their metabolites play important roles in a variety of biological processes. Several signal molecules are localized in a glycolipid-enriched microdomain on the cell surface, and their signals are regulated by the glycolipid composition. However, the function of glycolipids
W I Weis et al.
Annual review of biochemistry, 65, 441-473 (1996-01-01)
Lectins are responsible for cell surface sugar recognition in bacteria, animals, and plants. Examples include bacterial toxins; animal receptors that mediate cell-cell interactions, uptake of glycoconjugates, and pathogen neutralization; and plant toxins and mitogens. The structural basis for selective sugar
S Hakomori et al.
Journal of biochemistry, 118(6), 1091-1103 (1995-12-01)
Glycosphingolipids (GSLs), cell type-specific markers which change dramatically during ontogenesis and oncogenesis, have been implicated as playing major roles in cellular interactions and control of cell proliferation in multicellular organisms. These functional roles have been partially clarified through two types
Subroto Chatterjee et al.
Biochimica et biophysica acta, 1780(3), 370-382 (2007-12-14)
Although lactosylceramide (LacCer) plays a pivotal role in the biosynthesis of nearly all the major glycosphingolipids, its function in regulating cellular function has begun to emerge only recently. Our current opinion is that several physiologically critical molecules such as modified/oxidized
F Pincet et al.
Biophysical journal, 80(3), 1354-1358 (2001-02-27)
Carbohydrate-carbohydrate interactions are rarely considered in biologically relevant situations such as cell recognition and adhesion. One Ca(2+)-mediated homotypic interaction between two Lewis(x) determinants (Le(x)) has been proposed to drive cell adhesion in murine embryogenesis. Here, we confirm the existence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service