Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810221C

Avanti

C12-NBD Galactosyl Ceramide

Avanti Research - A Croda Brand 810221C

Synonym(s):

N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-D-galactosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C42H71N5O11
CAS Number:
Molecular Weight:
822.04
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810221C-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810221C

concentration

0.25 mg/mL (810221C-250ug)

shipped in

dry ice

storage temp.

−20°C

General description

Galactosyl Ceramide is a glycosphingolipid, predominantly present in myelin sheath.[1]

Biochem/physiol Actions

Abnormal accumulation of galactosyl ceramide in the central nervous system, due to failure in its degradation process results in globoid cell leukodystrophy.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810221C-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple Sclerosis: A Critical Conspectus, 195-195 (2012)
The Biosynthesis of Polysaccharides (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service