Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810220C

Avanti

C6-NBD Galactosyl Ceramide

Avanti Research - A Croda Brand 810220C

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-D-galactosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H59N5O11
CAS Number:
Molecular Weight:
737.88
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810220C-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810220C

concentration

0.25 mg/mL (810220C-250ug)

shipped in

dry ice

storage temp.

−20°C

General description

C6-NBD Galactosyl Ceramide is a fluorescent derivative of biologically active compound galactosyl ceramide. NBD (2-(4-nitro-2,1,3-benzoxadiazol-7-yl) aminoethyl) is a fluorophore. Galactosyl ceramide (Galactocerebroside) is produced in the lumen of the endoplasmic reticulum.

Application

C6-NBD Galactosyl Ceramide may be used as a substrate for neutral β -glycosylceramidase (GCase) activity assay.

Biochem/physiol Actions

Galactosyl ceramide (GC) is a glycosphingolipid and precursor of 3′-sulfo-galactosylceramide, a major sulfoglycolipid present in the brain and other organs. Galactosyl ceramide belongs to a new class of immunomodulators and vaccine adjuvants that are known to stimulate natural killer T (NKT) cells to increase secretion of T helper type 1 (Th1) and T-helper type 2 (Th2) cytokines. The excess accumulation of GC is related to diseased condition such as Krabbe′s disease.

Packaging

5 mL Amber Glass Screw Cap Vial (810220C-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent and Luminescent Probes for Biological Activity (1999)
Li Bai et al.
Journal of immunology (Baltimore, Md. : 1950), 188(7), 3053-3061 (2012-03-07)
α-Galactosylceramide represents a new class of vaccine adjuvants and immunomodulators that stimulate NKT cells to secrete Th1 and Th2 cytokines. Synthetic variants with short or unsaturated acyl chains exhibit a striking Th2 bias in vivo but no evidence of defect
Bernadette Breiden et al.
Annual review of biochemistry, 88, 461-485 (2019-06-22)
Glycosphingolipids are cell-type-specific components of the outer leaflet of mammalian plasma membranes. Gangliosides, sialic acid-containing glycosphingolipids, are especially enriched on neuronal surfaces. As amphi-philic molecules, they comprise a hydrophilic oligosaccharide chain attached to a hydrophobic membrane anchor, ceramide. Whereas glycosphingolipid
Distinct APCs explain the cytokine bias of $\alpha$-galactosylceramide variants in vivo
Bai LC, et al.
Journal of immunology (Baltimore, Md. : 1950), 188(7), 3053-3061 (2012)
Lysosomal lipid storage diseases
Schulze H and Sandhoff K
Cold Spring Harbor Perspectives in Biology, 3(6), a004804-a004804 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service