Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810214P

Avanti

C6-NBD Phytosphingosine

Avanti Research - A Croda Brand 810214P, powder

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-phytosphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H51N5O7
CAS Number:
Molecular Weight:
593.76
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (810214P-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810214P

shipped in

dry ice

Storage temp.

−20°C

General description

C6-NBD Phytosphingosine is a fluorescent analog of the sphingoid compound, phytosphingosine with NBD (2-(4-nitro-2,1,3-benzoxadiazol-7-yl) aminoethyl) being the fluorescent group.[1][2] Phytosphingosine is specifically localised to tissues such as skin, kidney, and intestine in animals and decrease in its production may lead to disorders. Phytosphingosine (PHS) is a major a long-chain base in yeast and mammalian cells.[2][3]

Application

C6-NBD Phytosphingosine N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-phytosphingosine is suitable to study membrane fusion with the help of fForster resonance energy transfer (FRET).[4][5]

Biochem/physiol Actions

Phytosphingosine (PHS) is an important component of phytoceramides. PHS metabolism results in the production of sphingolipids and glycerophospholipids.[2][3]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810214P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent and Luminescent Probes for Biological Activity (1999)
Endotoxin-adsorbing macrophage-mimetic hybrid liposome for sepsis treatment
Jiang LL, et al.
Chemical Engineering Journal, 371(1), 15-25 (2019)
Hyun Kyung Choi et al.
Archives of dermatological research, 309(10), 795-803 (2017-09-25)
Phytosphingosine (PHS) is a sphingoid that is a key component of phytoceramides NP, AP and EOP. PHS has been known to have anti-inflammation and antimicrobial activities and to stimulate epidermal differentiation. In addition, it is reported that PHS treatment notably
Chunai Gong et al.
Journal of nanobiotechnology, 18(1), 92-92 (2020-06-18)
Cell membrane- covered drug-delivery nanoplatforms have been garnering attention because of their enhanced bio-interfacing capabilities that originate from source cells. In this top-down technique, nanoparticles (NPs) are covered by various membrane coatings, including membranes from specialized cells or hybrid membranes
Natsuki Kondo et al.
Nature communications, 5, 5338-5338 (2014-10-28)
The long-chain base phytosphingosine is a component of sphingolipids and exists in yeast, plants and some mammalian tissues. Phytosphingosine is unique in that it possesses an additional hydroxyl group compared with other long-chain bases. However, its metabolism is unknown. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service