Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810214C

Avanti

C6-NBD Phytosphingosine

Avanti Research - A Croda Brand 810214C

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-phytosphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H51N5O7
CAS Number:
Molecular Weight:
593.76
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810214C-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810214C

concentration

0.05 mg/mL (810214C-50ug)

shipped in

dry ice

storage temp.

−20°C

General description

C6-NBD Phytosphingosine is a fluorescent derivative of phytosphingosine (PHS). Its fluorescent property is due to presence of NBD (2-(4-nitro-2,1,3-benzoxadiazol-7-yl) aminoethyl) group.[1]

Application

C6-NBD Phytosphingosine or N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-phytosphingosine is suitable for use in membrane fusion and verification studies using forster resonance energy transfer (FRET).[2][3]

Biochem/physiol Actions

Phytosphingosine (PHS) is a long-chain base sphingoid and a major constituent of various phytoceramides. PHS is abundantly present in skin, intestine, and kidneys of animals. It exhibits anti-inflammatory and antimicrobial properties. PHS is known to increase the expression of genes including transglutaminase 1 and involucrin, that are involved in keratinocyte differentiation in humans.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (810214C-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent and Luminescent Probes for Biological Activity null
Hyun Kyung Choi et al.
Archives of dermatological research, 309(10), 795-803 (2017-09-25)
Phytosphingosine (PHS) is a sphingoid that is a key component of phytoceramides NP, AP and EOP. PHS has been known to have anti-inflammation and antimicrobial activities and to stimulate epidermal differentiation. In addition, it is reported that PHS treatment notably
Endotoxin-adsorbing macrophage-mimetic hybrid liposome for sepsis treatment
Jiang LL, et al.
Chemical Engineering Journal, 371(1), 15-25 (2019)
Chunai Gong et al.
Journal of nanobiotechnology, 18(1), 92-92 (2020-06-18)
Cell membrane- covered drug-delivery nanoplatforms have been garnering attention because of their enhanced bio-interfacing capabilities that originate from source cells. In this top-down technique, nanoparticles (NPs) are covered by various membrane coatings, including membranes from specialized cells or hybrid membranes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service