Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810212P

Avanti

C6-NBD Sphinganine

Avanti Research - A Croda Brand 810212P, powder

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H51N5O6
CAS Number:
Molecular Weight:
577.76
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (810212P-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810212P

shipped in

dry ice

storage temp.

−20°C

General description

C6-Sphinganine is an analogue of sphinganine (dihydrosphingosine), with C6 being the number of carbon atoms in the acyl residue.[1] C6-NBD Sphinganine is an omega fatty acid labeled fluorescent sphingolipid with 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) as the fluorophore.

Packaging

5 mL Amber Glass Screw Cap Vial (810212P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Thevissen et al.
Bioorganic & medicinal chemistry letters, 18(13), 3728-3730 (2008-06-06)
The minimal fungicidal concentration (MFC) of dihydrosphingosine (DHS), phytosphingosine (PHS), and five short-chain DHS derivatives was determined for Candida albicans and Candida glabrata. In this respect, a C15- and a C17-homologue of DHS showed a 2- to 10-fold decreased MFC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service