Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810206X

Avanti

NBD Sphinganine

omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol, chloroform:methanol (9:1)

Synonym(s):

D-erythro-sphinganine-N-NBD; NBD-dihydrosphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H41N5O5
CAS Number:
Molecular Weight:
479.61
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810206X-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810206X

concentration

0.25 mg/mL (810206X-250ug)

shipped in

dry ice

storage temp.

−20°C

General description

Sphinganine (dihydrosphingosine) is a common sphingoid base in phospholipids found in lesser amounts compared to sphingosine.[1]

Application

NBD Sphinganine or omega (7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol is suitable for the determination of the dihydrosphingosine (DHS) localization to endoplasmic reticulum for the activation of activating transcription factor 6 (ATF6) protein.[2]

Packaging

5 mL PTFE Screw Cap Vial with Foil Bag Argon overseal (810206X-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Liver,Kidney, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arvin B Tam et al.
Developmental cell, 46(3), 327-343 (2018-08-08)
The unfolded protein response (UPR) is induced by proteotoxic stress of the endoplasmic reticulum (ER). Here we report that ATF6, a major mammalian UPR sensor, is also activated by specific sphingolipids, dihydrosphingosine (DHS) and dihydroceramide (DHC). Single mutations in a previously
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service