810193P
Avanti
16:0-12:0 NBD PS
Avanti Research™ - A Croda Brand 810193P, powder
Synonym(s):
1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoserine (ammonium salt)
Select a Size
Select a Size
About This Item
assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (810193P-1mg)
pkg of 5 × 1 mg (810193P-5mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 810193P
shipped in
dry ice
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Packaging
Legal Information
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service