Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810175C

Avanti

18:1-06:0 NBD PA

Avanti Research - A Croda Brand 810175C

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H56N5O11P
CAS Number:
Molecular Weight:
729.80
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810175C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810175C

concentration

1 mg/mL (810175C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is an amphipathic lipid containing a polar head group and two long hydrophobic acyl chains.[1] PA is an anionic and a membrane structural lipid.[2] 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) PA is a fluorescent analog NBD attached to the sn-2 fatty acid of PA.[3]

Application

18:1-06:0 NBD PA or 1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphate (ammonium salt) is suitable to study the effects of NBD-PA induced vesiculation on epidermal growth factor receptor (EGFR) endocytosis in mammalian cells [4] and the co-localization of fluorescently-labeled Poly(A)-Specific Ribonuclease (PARN) with the NBD-tagged PA that outlines the close proximity phospholipase D (PLD2) and PARN.[5]

Biochem/physiol Actions

Phosphatidic acid (PA) is a secondary messenger involved in various cellular processes and a bioactive lipid.[6] It acts as a parent compound for more complex membrane phospholipids.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810175C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive Lipids
An Introduction to Biological Membranes, , number=(, pages=), 453-478 (2016)
Lipid second messengers and Receptors
Encyclopedia of Endocrine Diseases, 77(3), 329-334 (2004)
Taylor E Miller et al.
Biology open, 6(2), 176-186 (2016-12-25)
The removal of mRNA transcript poly(A) tails by 3'→5' exonucleases is the rate-limiting step in mRNA decay in eukaryotes. Known cellular deadenylases are the CCR4-NOT and PAN complexes, and poly(A)-specific ribonuclease (PARN). The physiological roles and regulation for PARN is
Priya Putta et al.
Biochimica et biophysica acta, 1858(11), 2709-2716 (2016-08-03)
Phosphatidic acid (PA) is a crucial membrane phospholipid involved in de novo lipid synthesis and numerous intracellular signaling cascades. The signaling function of PA is mediated by peripheral membrane proteins that specifically recognize PA. While numerous PA-binding proteins are known
Phosphatidic acid binding proteins display differential binding as a function of membrane curvature stress and chemical properties
Putta P, et al.
Biochimica et Biophysica Acta, 1858(11), 2709-2716 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service