Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810168P

Avanti

06:0-12:0 NBD PG

Avanti Research - A Croda Brand 810168P, powder

Synonym(s):

1-hexanoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52N5O13P
CAS Number:
Molecular Weight:
721.73
UNSPSC Code:
12352211

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810168P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810168P

shipped in

dry ice

Storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (810168P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service