Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810166C

Avanti

18:1-12:0 NBD PG

Avanti Research - A Croda Brand 810166C

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C42H74N5O13P
CAS Number:
Molecular Weight:
888.04
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 5 × 1 mL (810166C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810166C

concentration

1 mg/mL (810166C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylglycerol (PG) is an anionic phospholipid[1], constituting 20−25% in E. coli membranes.[2] It synthesized by phosphatidic acid.[3] Nitrobenzoxadiazole phosphoglycerol (NBD PG) is a fluorescently labeled phospholipid with NBD attached to the fatty acid chain of the phosphatidylglycerol.[2]

Application

18:1-12:0 NBD PG or 1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt) has been used in the lipid transfection experiment for the analysis of tissue-specific changes in lipid utilization.[4]

Biochem/physiol Actions

Phosphatidylglycerol is involved in the biosynthesis of cardiolipin.[1] It also contributes to the photosynthetic activity by maintaining the structural photosynthetic apparatus.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810166C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Evidence for Requirement of Phosphatidylglycerol in Photosystem II of Photosynthesis
Hagio M, et al.
Plant Pathology null
Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Plant & Cell Physiology null
Specificity of the transport of lipid II by FtsW in Escherichia coli
Mohammadi T
Journal of Biochemistry and Molecular Biology Research null
A nutritional memory effect counteracts the benefits of dietary restriction in old mice
Hahn O, et al.
Nature Biomedical Engineering null
Meijing Wang et al.
Biochimica et biophysica acta, 1831(6), 1089-1097 (2013-02-06)
Gold nanoparticles (GNPs) have been applied as diagnostic and therapeutic agents because they can be targeted, localized, and be heated to cause cell death. However, their use has been limited by their relatively low biocompatibility. In this work, we coated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service