Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810165C

Avanti

18:1-06:0 NBD PG

Avanti Research - A Croda Brand 810165C

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62N5O13P
CAS Number:
Molecular Weight:
803.88
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810165C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810165C

concentration

1 mg/mL (810165C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylglycerol (PG) is an anionic phospholipid and is a predominant component of the cell membrane. Nitrobenzoxadiazole phosphoglycerol (NBD PG) is a fluorophore-labeled phospholipid where 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) is linked to the fatty acid chain of the phosphatidylglycerol.

Application

18:1-06:0 NBD PG may be used in the determination of phosphatidylglycerol distribution across the bilayer of the membrane and to measure the rate constant for transbilayer movement of lipid molecules by preparing giant unilamellar vesicles (GUVs).

Biochem/physiol Actions

Phosphatidylglycerol contributes to the biosynthesis of cardiolipin. It plays a vital role in photosynthetic activity by maintaining the structural photosynthetic apparatus.

Packaging

5 mL Amber Glass Screw Cap Vial (810165C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of membrane tension on transbilayer movement of lipids
Hasan M, et al.
J. Chem. Phys. null
Specificity of the transport of lipid II by FtsW in Escherichia coli
Mohammadi T, et al.
Journal of Biochemistry and Molecular Biology Research null
Direct Evidence for Requirement of Phosphatidylglycerol in Photosystem II of Photosynthesis
Hagio M, et al.
Plant Physiology null
Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Plant & Cell Physiology null
Impact of the ?-Lactam Resistance Modifier (
Rosado H, et al.
International Journal of Molecular Sciences null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service