Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810164C

Avanti

16:0-12:0 NBD PG

Avanti Research - A Croda Brand 810164C

Synonym(s):

1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H72N5O13P
CAS Number:
Molecular Weight:
862.00
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810164C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810164C

concentration

1 mg/mL (810164C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

16:0-12:0 NBD PG is a fluorophore-labeled phospholipid.[1]
Phosphatidylglycerol (PG), an anionic phospholipid is a predominant component of the cell membrane.[2]

Application

16:0-12:0 NBD PG has been used in microfluidic chip-based technology to demonstrate separation of fluorophore-labeled phospholipids.[1]
16:0-12:0 NBD PG or 1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt) is suitable for the separation of fluorophore-labeled phospholipids in microfluidic chip-based technology.[3]

Biochem/physiol Actions

Phosphatidylglycerol contributes to photosynthetic activity by maintaining the structural photosynthetic apparatus.[4] It is useful for the biosynthesis of cardiolipin.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810164C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of phospholipids in microfluidic chip device: application to high-throughput screening assays for lipid-modifying enzymes
Lin S, et al.
Analytical Biochemistry null
Phosphatidylglycerol is Essential for the Development of Thylakoid Membranes in Arabidopsis thaliana
Hagio M, et al.
Plant & Cell Physiology null
Direct Evidence for Requirement of Phosphatidylglycerol in Photosystem II of Photosynthesis
Hagio M, et al.
Plant Physiology null
Separation of phospholipids in microfluidic chip device: Application to high-throughput screening assays for lipid-modifying enzymes
Lin S, et al.
Analytical Biochemistry, 314(1), 97-107 (2003)
Furong Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 36(11), 2946-2953 (2020-02-26)
Phospholipase A2 is an important enzyme species which can widely be found in animals, plants, bacteria, and so on. A large number of studies have shown that phospholipase A2 is highly catalytic toward the lipids. Here, sum frequency generation (SFG)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service