Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810159C

Avanti

18:1 NBD-PEG4 Lyso PE

Avanti Research - A Croda Brand 810159C

Synonym(s):

1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine-N-[tetra(ethylene glycol)]-N′-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H71N6O15P
CAS Number:
Molecular Weight:
907.00
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810159C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810159C

concentration

1 mg/mL (810159C-1mg)

shipped in

dry ice

storage temp.

−20°C

Biochem/physiol Actions

Fluorescent labelled lipids are generally useful in cell biology and biophysical studies. Cellular events involving lipids such as lipid domain generation, disaggregation and re-organization can be studied with fluorescent lipids. They are also useful in analyzing the physical properties of lipid bilayers. Nitrobenzoxadiazole (NBD)-labeled lipids are commonly applicable as fluorescent membrane probes. They exhibit a good quantum yield in lipid bilayers. NBD-lipids can be used for sensing slow solvent relaxation by the red-edge excitation shift (REES) phenomenon.

Packaging

5 mL Amber Glass Screw Cap Vial (810159C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerstin Pinkwart et al.
The Journal of biological chemistry, 294(34), 12599-12609 (2019-07-05)
Cholesterol constitutes ∼30-40% of the mammalian plasma membrane, a larger fraction than of any other single component. It is a major player in numerous signaling processes as well as in shaping molecular membrane architecture. However, our knowledge of the dynamics
Christian Kleusch et al.
Molecules (Basel, Switzerland), 17(1), 1055-1073 (2012-01-24)
In this paper a rapid and highly efficient method for controlled incorporation of fluorescent lipids into living mammalian cells is introduced. Here, the fluorescent molecules have two consecutive functions: First, they trigger rapid membrane fusion between cellular plasma membranes and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service