Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810152C

Avanti

14:0-12:0 NBD PE

Avanti Research - A Croda Brand 810152C

Synonym(s):

1-myristoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H64N5O11P
CAS Number:
Molecular Weight:
785.90
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810152C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810152C

concentration

1 mg/mL (810152C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Nitrobenzoxadiazole phosphoethanolamine (NBD PE) is a fluorescent probe with NBD covalently attached to the headgroup of phosphatidylethanolamine.[1] It localized at the membrane interface[2] and mostly present in brain.[3] Phosphoethanolamine is the second most abundant, metabolically related aminophospholipid.[3]

Biochem/physiol Actions

Phosphoethanolamine of cardiac myocytes is useful in the prevention of cell damage after ischemia. Nitrobenzoxadiazole phosphoethanolamine (NBD PE) is used to monitor the Red edge excitation shift (REES) effects due to its unique motional and dielectric properties.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810152C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of NBD-labeled lipids in membrane and cell biology
Fluorescent methods to study biological membranes, 37-50 (2012)
Jean E Vance et al.
Biochimica et biophysica acta, 1831(3), 543-554 (2012-09-11)
Phosphatidylserine (PS) and phosphatidylethanolamine (PE) are metabolically related membrane aminophospholipids. In mammalian cells, PS is required for targeting and function of several intracellular signaling proteins. Moreover, PS is asymmetrically distributed in the plasma membrane. Although PS is highly enriched in
S C Lopes et al.
Biochimica et biophysica acta. Biomembranes, 1861(6), 1152-1161 (2019-03-07)
Mitochondrial membranes are pointed out as the site of cardiotoxic action of local anaesthetics. Its three main phospholipids components are phosphatidylcholine, phosphatidylethanolamine and cardiolipin. Cardiolipins, in eukaryotes, are only found in mitochondria and are essential for the maintenance of its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service