Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810135P

Avanti

Acyl 12:0 NBD PE

Avanti Research - A Croda Brand 810135P, powder

Synonym(s):

1-acyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68N5O11P
CAS Number:
Molecular Weight:
813.96
UNSPSC Code:
12352211

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810135P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810135P

shipped in

dry ice

Storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (810135P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service