Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810110C

Avanti

12-NBD Stearate

Avanti Research - A Croda Brand 810110C

Synonym(s):

12-N-methyl-(7-nitrobenz-2-oxa-1,3-diazo)aminostearic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H40N4O5
CAS Number:
Molecular Weight:
476.61
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810110C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810110C

concentration

1 mg/mL (810110C-1mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C25H40N4O5/c1-3-4-5-12-15-20(16-13-10-8-6-7-9-11-14-17-23(30)31)28(2)21-18-19-22(29(32)33)25-24(21)26-34-27-25/h18-20H,3-17H2,1-2H3,(H,30,31)

InChI key

SAAVELCIPHKGJE-UHFFFAOYSA-N

General description

12-NBD Stearate is a fluorescent long-chain fatty acid (LCFA) derivative.[1] It is amphipathic and a non-esterifiable fatty acid probe for incorporation into lipid droplets.[2]

Application

12-NBD Stearate may be used:
  • as a ligand for fatty-acid binding protein 5 (FABP5) in competitive binding based high throughput fluorescence displacement assay[3]
  • as a ligand for FABPs in spectrofluorometric measurements[4]
  • as an inhibitor for FABP3, FABP5 and FABP7[5]

Biochem/physiol Actions

12-NBD Stearate is unmetabolizable in HepG2 cells and colocalizes with α-caveolin-1[1] and its uptake is inhibited by filipin III.[1] The sterol carrier protein-2 (SCP-2) 15-kDa isoform favors NBD Stearate transport compared to its 13.2-kDa isoform.[2] The fatty-acid binding protein (FABP) expression in differentiating embryonic stem cells (ES) regulates the uptake of NBD Stearate.[6] NBD Stearate is used majorly in fluorescence displacement binding assays of fatty-acid binding proteins.[7]

Packaging

5 mL Amber Glass Screw Cap Vial (810110C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of fatty acid binding proteins elevates brain anandamide levels and produces analgesia
Kaczocha M, et al.
Testing, 9(4) (2014)
Fatty acid-binding proteins transport N-acylethanolamines to nuclear receptors and are targets of endocannabinoid transport inhibitors
Kaczocha M, et al.
The Journal of Biological Chemistry, 287(5), 3415-3424 (2012)
Targeting fatty acid binding protein (FABP) anandamide transporters-a novel strategy for development of anti-inflammatory and anti-nociceptive drugs
Berger WT, et al.
Testing, 7(12) (2012)
FABP1: a novel hepatic endocannabinoid and cannabinoid binding protein
Huang H, et al.
Biochemistry, 55(37), 5243-5255 (2016)
B P Atshaves et al.
The Journal of biological chemistry, 276(27), 25324-25335 (2001-05-03)
Despite the critical role lipid droplets play in maintaining energy reserves and lipid stores for the cell, little is known about the regulation of the lipid or protein components within the lipid droplet. Although immunofluorescence of intact cells as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service